Product Details

SNP ID
rs147061228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53460995 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGAACTGTTTTTCTCTGATTTTG[A/G]ATTTCTTTTTACTCCAAAGAGTACA
Phenotype
MIM: 601210
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCBP2 PubMed Links

Gene Details

Gene
PCBP2
Gene Name
poly(rC) binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098620.2 Intron NP_001092090.1
NM_001128911.1 Intron NP_001122383.1
NM_001128912.1 Intron NP_001122384.1
NM_001128913.1 Intron NP_001122385.1
NM_001128914.1 Intron NP_001122386.1
NM_005016.5 Intron NP_005007.2
NM_031989.4 Intron NP_114366.1
Gene
PCBP2-OT1
Gene Name
PCBP2 overlapping transcript 1
There are no transcripts associated with this gene.

View Full Product Details