Product Details

SNP ID
rs147230079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:76025461 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTCCACAGGTCGGTTGTCTCAAT[A/G]GATAACTTTATTTGAAATGAAATGC
Phenotype
MIM: 605335
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHLDA1 PubMed Links

Gene Details

Gene
PHLDA1
Gene Name
pleckstrin homology like domain family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007350.3 5899 UTR 3 NP_031376.3

View Full Product Details