Product Details

SNP ID
rs147252656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:9150734 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAACCATGAAGGAAAAACTTAGCG[C/T]CTGATTTGTCACCTGAAAGGAAAAG
Phenotype
MIM: 604874 MIM: 176420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLRG1 PubMed Links

Gene Details

Gene
KLRG1
Gene Name
killer cell lectin like receptor G1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005810.3 4319 Intron NP_005801.3
XM_017018682.1 4319 Intron XP_016874171.1
XM_017018683.1 4319 Intron XP_016874172.1
XM_017018684.1 4319 Intron XP_016874173.1
XM_017018685.1 4319 Intron XP_016874174.1
Gene
PZP
Gene Name
PZP, alpha-2-macroglobulin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002864.2 4319 Missense Mutation ACG,GCG T1432A NP_002855.2
XM_011520804.2 4319 Missense Mutation ACG,GCG T1432A XP_011519106.1
XM_011520805.2 4319 Missense Mutation ACG,GCG T1431A XP_011519107.1
XM_011520806.1 4319 Intron XP_011519108.1
XM_011520807.2 4319 Intron XP_011519109.1
XM_017019763.1 4319 Missense Mutation ACG,GCG T1432A XP_016875252.1
XM_017019764.1 4319 Missense Mutation ACG,GCG T1432A XP_016875253.1
XM_017019765.1 4319 Missense Mutation ACG,GCG T1349A XP_016875254.1
XM_017019766.1 4319 Missense Mutation ACG,GCG T1218A XP_016875255.1
XM_017019767.1 4319 Intron XP_016875256.1

View Full Product Details