Product Details

SNP ID
rs147703598
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:50925177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTTGGTGAGGTTCACTGTGATA[C/T]ACAACGAACAGATGGCAAGCAAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
METTL7A PubMed Links

Gene Details

Gene
METTL7A
Gene Name
methyltransferase like 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014033.3 427 Missense Mutation CAC,TAC H47Y NP_054752.3
XM_006719332.1 427 Missense Mutation CAC,TAC H47Y XP_006719395.1

View Full Product Details