Product Details

SNP ID
rs148009650
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132771791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTTGTTACCGCCTGCGACCTAC[A/G]CCTGGCTGACTCCAGGGGAAACCGC
Phenotype
MIM: 616062 MIM: 602581
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKLE2 PubMed Links

Gene Details

Gene
ANKLE2
Gene Name
ankyrin repeat and LEM domain containing 2
There are no transcripts associated with this gene.

Gene
GOLGA3
Gene Name
golgin A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172557.1 3975 Intron NP_001166028.1
NM_005895.3 3975 UTR 3 NP_005886.2
XM_005266164.4 3975 UTR 3 XP_005266221.1
XM_005266167.4 3975 UTR 3 XP_005266224.1
XM_006719736.3 3975 UTR 3 XP_006719799.1
XM_006719737.3 3975 UTR 3 XP_006719800.1
XM_011534793.2 3975 UTR 3 XP_011533095.1
XM_011534794.2 3975 UTR 3 XP_011533096.1

View Full Product Details