Product Details

SNP ID
rs148375896
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7095576 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCCATTAGGAAAGTGTGAGCTA[A/G]AGGATACCATTTTCGCAGAAGCAGG
Phenotype
MIM: 613785 MIM: 608974
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1R PubMed Links

Gene Details

Gene
C1R
Gene Name
complement C1r subcomponent
There are no transcripts associated with this gene.

Gene
C1RL
Gene Name
complement C1r subcomponent like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297640.1 2424 UTR 3 NP_001284569.1
NM_001297642.1 2424 UTR 3 NP_001284571.1
NM_001297643.1 2424 Intron NP_001284572.1
NM_016546.3 2424 UTR 3 NP_057630.2
XM_017019397.1 2424 UTR 3 XP_016874886.1
XM_017019398.1 2424 UTR 3 XP_016874887.1
XM_017019399.1 2424 Intron XP_016874888.1
XM_017019400.1 2424 Intron XP_016874889.1
XM_017019401.1 2424 Intron XP_016874890.1
XM_017019402.1 2424 Intron XP_016874891.1
XM_017019403.1 2424 Intron XP_016874892.1
XM_017019404.1 2424 Intron XP_016874893.1

View Full Product Details