Product Details

SNP ID
rs148697931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53065128 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAAGGAGTGAAGGCCCTAGTGC[C/T]CTGTCACCTCACAGCCCCTCTCTCT
Phenotype
MIM: 607717
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR6757 PubMed Links

Gene Details

Gene
MIR6757
Gene Name
microRNA 6757
There are no transcripts associated with this gene.

Gene
SPRYD3
Gene Name
SPRY domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032840.2 2120 UTR 3 NP_116229.1
Gene
TNS2
Gene Name
tensin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015319.2 2120 Intron NP_056134.2
NM_170754.2 2120 Intron NP_736610.2
NM_198316.1 2120 Intron NP_938072.1
XM_006719302.3 2120 Intron XP_006719365.1
XM_006719303.1 2120 Intron XP_006719366.1
XM_011538079.1 2120 Intron XP_011536381.1
XM_017019088.1 2120 Intron XP_016874577.1
XM_017019089.1 2120 Intron XP_016874578.1

View Full Product Details