Product Details

SNP ID
rs148967345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:85018479 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAATTTCACTAATAGTAGTGATAT[A/G]ATAGACAGAATTGTTTCCTCATCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TSPAN19 PubMed Links

Gene Details

Gene
TSPAN19
Gene Name
tetraspanin 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100917.1 Intron NP_001094387.1
XM_005268669.4 Intron XP_005268726.1
XM_017018862.1 Intron XP_016874351.1
XM_017018863.1 Intron XP_016874352.1
XM_017018864.1 Intron XP_016874353.1

View Full Product Details