Product Details

SNP ID
rs149519721
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:81078328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACAAGACCCACTTCGCAGCCTCG[A/G]TGACCGACCCCGAGAGGTTCTGGGG
Phenotype
MIM: 614356
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSS3 PubMed Links

Gene Details

Gene
ACSS3
Gene Name
acyl-CoA synthetase short-chain family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005269150.2 311 Missense Mutation ATG,GTG M70V XP_005269207.1
XM_005269151.4 311 Missense Mutation ATG,GTG M70V XP_005269208.1
XM_011538740.2 311 Intron XP_011537042.1

View Full Product Details