Product Details

SNP ID
rs150059138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:93678817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAGTACTCCGCTCACTTCGCCTG[A/G]AGCTGGGTGCAGAGGTATTGGTGGA
Phenotype
MIM: 603454
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CRADD PubMed Links

Gene Details

Gene
CRADD
Gene Name
CASP2 and RIPK1 domain containing adaptor with death domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320099.1 174 Missense Mutation AAG,GAG K15E NP_001307028.1
NM_001320100.1 174 Missense Mutation AAG,GAG K15E NP_001307029.1
NM_001320101.1 174 Missense Mutation AAG,GAG K15E NP_001307030.1
NM_003805.4 174 Missense Mutation AAG,GAG K15E NP_003796.1
XM_017020142.1 174 Missense Mutation AAG,GAG K15E XP_016875631.1
XM_017020143.1 174 Missense Mutation AAG,GAG K15E XP_016875632.1
XM_017020144.1 174 Missense Mutation AAG,GAG K15E XP_016875633.1
XM_017020145.1 174 Missense Mutation AAG,GAG K15E XP_016875634.1

View Full Product Details