Product Details

SNP ID
rs150119995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:56339817 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCAGCTTCATGCCTCCCTACTG[A/G]GCCTCAGCCAACTCCTGCAGGTATG
Phenotype
MIM: 605580 MIM: 600556
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL23A PubMed Links

Gene Details

Gene
IL23A
Gene Name
interleukin 23 subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016584.2 741 Missense Mutation AGC,GGC S130G NP_057668.1
XM_011538477.2 741 Missense Mutation AGC,GGC S130G XP_011536779.1
Gene
PAN2
Gene Name
PAN2 poly(A) specific ribonuclease subunit
There are no transcripts associated with this gene.

Gene
STAT2
Gene Name
signal transducer and activator of transcription 2
There are no transcripts associated with this gene.

View Full Product Details