Product Details

SNP ID
rs150952812
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:76026498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGAGAAATACAGAGTGTTTGGTA[C/T]ATGAGAGAAAAAAAGTTAAAACAGG
Phenotype
MIM: 605335
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHLDA1 PubMed Links

Gene Details

Gene
PHLDA1
Gene Name
pleckstrin homology like domain family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007350.3 4862 UTR 3 NP_031376.3

View Full Product Details