Product Details

SNP ID
rs151159720
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55365730 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTAGCTGTGGTAACACTGATGG[C/T]CACCTTGACATTAGTTATTCTCTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR6C75 PubMed Links

Gene Details

Gene
OR6C75
Gene Name
olfactory receptor family 6 subfamily C member 75
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005497.1 620 Missense Mutation GCC,GTC A207V NP_001005497.1

View Full Product Details