Product Details
- SNP ID
-
rs137869269
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:49669783 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACGTTTTTAATTACTTACATAAAG[A/C]AGCAATCAAGCCATCGGAATTTGCA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EBPL
PubMed Links
Gene Details
- Gene
- EBPL
- Gene Name
- emopamil binding protein like
View Full Product Details