Product Details

SNP ID
rs138495499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:35772655 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTGATTTTGCGTGTTATCACTTC[C/T]GGATGCTCTAAAAGGGGGGGAAAAA
Phenotype
MIM: 604742
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCLK1 PubMed Links

Gene Details

Gene
DCLK1
Gene Name
doublecortin like kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195415.1 3589 UTR 3 NP_001182344.1
NM_001195416.1 3589 UTR 3 NP_001182345.1
NM_001195430.1 3589 Intron NP_001182359.1
NM_004734.4 3589 UTR 3 NP_004725.1
XM_005266592.2 3589 UTR 3 XP_005266649.1
XM_006719893.2 3589 UTR 3 XP_006719956.1
XM_017020847.1 3589 UTR 3 XP_016876336.1
XM_017020848.1 3589 Intron XP_016876337.1

View Full Product Details