Product Details

SNP ID
rs138892103
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39012133 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCATTATGCTGTAATTCTTGCA[A/C]GGATGATTGTGAAGGATTCTGAGGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PROSER1 PubMed Links

Gene Details

Gene
PROSER1
Gene Name
proline and serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025138.4 3469 Missense Mutation GTG,TTG V888L NP_079414.3
XM_005266545.3 3469 Missense Mutation GTG,TTG V866L XP_005266602.1
XM_011535239.2 3469 Missense Mutation GTG,TTG V859L XP_011533541.1

View Full Product Details