Product Details

SNP ID
rs140518706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110879669 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGGTGCAGGTGCATGGTGCCCA[A/G]CACGGCACTGTCGTACCGCTCCTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD10 PubMed Links

Gene Details

Gene
ANKRD10
Gene Name
ankyrin repeat domain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286721.1 1404 Intron NP_001273650.1
NM_017664.3 1404 Silent Mutation CTG,TTG L411L NP_060134.2
Gene
LINC00346
Gene Name
long intergenic non-protein coding RNA 346
There are no transcripts associated with this gene.

View Full Product Details