Product Details

SNP ID
rs142237911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:28700776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTTCCCAATTACCCATTACATA[C/T]AGAAATATTATCATGCCTGTCACCG
Phenotype
MIM: 616764
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC46A3 PubMed Links

Gene Details

Gene
SLC46A3
Gene Name
solute carrier family 46 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135919.1 2105 UTR 3 NP_001129391.1
NM_181785.3 2105 UTR 3 NP_861450.1
XM_005266361.2 2105 UTR 3 XP_005266418.1

View Full Product Details