Product Details

SNP ID
rs143687334
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:39012667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTTAGGTGAAGAATAATTTTAT[C/T]CTATTTCCAAACTATCCACATACCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PROSER1 PubMed Links

Gene Details

Gene
PROSER1
Gene Name
proline and serine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025138.4 Intron NP_079414.3
XM_005266545.3 Intron XP_005266602.1
XM_011535239.2 Intron XP_011533541.1

View Full Product Details