Product Details
- SNP ID
-
rs143872712
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:110881259 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAAGAGCTCAGTTCTAATAACTTT[G/T]AGAATGAGGACGCAGCTTAAATACA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD10
PubMed Links
Gene Details
- Gene
- ANKRD10
- Gene Name
- ankyrin repeat domain 10
View Full Product Details