Product Details

SNP ID
rs144018046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:49255885 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCCTTAGGAGATAATGAAGAGA[A/G]GACCAGAGTATCTACTGACAAAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDADC1 PubMed Links

Gene Details

Gene
CDADC1
Gene Name
cytidine and dCMP deaminase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193478.1 351 Missense Mutation AAG,AGG K75R NP_001180407.1
NM_030911.3 351 Missense Mutation AAG,AGG K75R NP_112173.1
XM_006719871.3 351 Intron XP_006719934.1
XM_011535250.2 351 Missense Mutation AAG,AGG K89R XP_011533552.1
XM_011535251.1 351 Missense Mutation AAG,AGG K30R XP_011533553.1
XM_011535253.1 351 Missense Mutation AGA,GGA R4G XP_011533555.1
XM_017020778.1 351 UTR 5 XP_016876267.1
XM_017020779.1 351 UTR 5 XP_016876268.1
XM_017020780.1 351 UTR 5 XP_016876269.1
XM_017020781.1 351 UTR 5 XP_016876270.1
XM_017020782.1 351 UTR 5 XP_016876271.1
XM_017020783.1 351 UTR 5 XP_016876272.1
XM_017020784.1 351 Intron XP_016876273.1
Gene
LOC105370203
Gene Name
uncharacterized LOC105370203
There are no transcripts associated with this gene.

View Full Product Details