Product Details

SNP ID
rs144126246
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:53029111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGCTTTTGGGTACCTGAATACA[A/G]TTTCATGGATGGCTAGACCTGGGCA
Phenotype
MIM: 614061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OLFM4 PubMed Links

Gene Details

Gene
OLFM4
Gene Name
olfactomedin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006418.4 Intron NP_006409.3

View Full Product Details