Product Details

SNP ID
rs144776457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:94439720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCACTTTAGTTGGGTTTGTTAAA[C/T]AAGCAAGCAAAGCGGAAACTACAGC
Phenotype
MIM: 191275
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DCT PubMed Links

Gene Details

Gene
DCT
Gene Name
dopachrome tautomerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129889.2 2165 UTR 3 NP_001123361.1
NM_001322182.1 2165 UTR 3 NP_001309111.1
NM_001322183.1 2165 UTR 3 NP_001309112.1
NM_001322184.1 2165 UTR 3 NP_001309113.1
NM_001322185.1 2165 UTR 3 NP_001309114.1
NM_001322186.1 2165 UTR 3 NP_001309115.1
NM_001922.4 2165 UTR 3 NP_001913.2
XM_011521049.1 2165 UTR 3 XP_011519351.1
XM_017020401.1 2165 UTR 3 XP_016875890.1

View Full Product Details