Product Details

SNP ID
rs145613367
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:110879766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTTTCAGCAGTGTCCCCAAACCC[A/G]TGGTAGTGTCCATAGTACAGGTTAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANKRD10 PubMed Links

Gene Details

Gene
ANKRD10
Gene Name
ankyrin repeat domain 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286721.1 1307 Intron NP_001273650.1
NM_017664.3 1307 Silent Mutation CAC,CAT H378H NP_060134.2
Gene
LINC00346
Gene Name
long intergenic non-protein coding RNA 346
There are no transcripts associated with this gene.

View Full Product Details