Product Details

SNP ID
rs147553289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:102786372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAACAATATAAAAGCATTCCTTA[C/T]AAAACACTTTATATCCACCCAATGA
Phenotype
MIM: 611613
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDELC1 PubMed Links

Gene Details

Gene
KDELC1
Gene Name
KDEL motif containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318732.1 Intron NP_001305661.1
NM_024089.2 Intron NP_076994.2
XM_005254075.2 Intron XP_005254132.1

View Full Product Details