Product Details

SNP ID
rs149952682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:79314023 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGCCTCCTGAGAATCTATGAAG[A/G]AATACTCAGAAACTAAGTGCAAACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RBM26 PubMed Links

Gene Details

Gene
RBM26
Gene Name
RNA binding motif protein 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286631.1 5622 Intron NP_001273560.1
NM_001286632.1 5622 Intron NP_001273561.1
NM_022118.4 5622 Intron NP_071401.3
XM_005266491.4 5622 UTR 3 XP_005266548.1
XM_005266497.1 5622 UTR 3 XP_005266554.1
XM_006719857.1 5622 UTR 3 XP_006719920.1
XM_011535179.1 5622 Intron XP_011533481.1
XM_011535180.2 5622 UTR 3 XP_011533482.1
XM_011535181.1 5622 Intron XP_011533483.1
XM_011535182.1 5622 Intron XP_011533484.1
XM_011535183.2 5622 UTR 3 XP_011533485.1
XM_011535184.1 5622 Intron XP_011533486.1
XM_011535185.2 5622 UTR 3 XP_011533487.1
XM_011535186.2 5622 Intron XP_011533488.1
XM_011535187.2 5622 UTR 3 XP_011533489.1
XM_011535188.2 5622 UTR 3 XP_011533490.1
XM_011535189.1 5622 Intron XP_011533491.1
XM_011535190.1 5622 Intron XP_011533492.1
XM_011535191.2 5622 UTR 3 XP_011533493.1
XM_011535192.2 5622 UTR 3 XP_011533494.1
XM_011535193.2 5622 UTR 3 XP_011533495.1
XM_011535195.1 5622 Intron XP_011533497.1
XM_017020688.1 5622 UTR 3 XP_016876177.1
XM_017020689.1 5622 Intron XP_016876178.1
XM_017020690.1 5622 UTR 3 XP_016876179.1
XM_017020691.1 5622 UTR 3 XP_016876180.1
XM_017020692.1 5622 UTR 3 XP_016876181.1
XM_017020693.1 5622 UTR 3 XP_016876182.1
XM_017020694.1 5622 UTR 3 XP_016876183.1
XM_017020695.1 5622 Intron XP_016876184.1
XM_017020696.1 5622 Intron XP_016876185.1
XM_017020697.1 5622 UTR 3 XP_016876186.1
XM_017020698.1 5622 Intron XP_016876187.1
XM_017020699.1 5622 UTR 3 XP_016876188.1
XM_017020700.1 5622 Intron XP_016876189.1
XM_017020701.1 5622 UTR 3 XP_016876190.1
XM_017020702.1 5622 UTR 3 XP_016876191.1
XM_017020703.1 5622 UTR 3 XP_016876192.1

View Full Product Details