Product Details

SNP ID
rs150399961
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:94440039 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCAGTGTTCCCATGACTACTAA[C/G]AGAGTTGTGGGCCAACCTGGAGTTT
Phenotype
MIM: 191275
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DCT PubMed Links

Gene Details

Gene
DCT
Gene Name
dopachrome tautomerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129889.2 1846 Silent Mutation CTC,CTG L506L NP_001123361.1
NM_001322182.1 1846 Silent Mutation CTC,CTG L162L NP_001309111.1
NM_001322183.1 1846 Silent Mutation CTC,CTG L162L NP_001309112.1
NM_001322184.1 1846 Silent Mutation CTC,CTG L162L NP_001309113.1
NM_001322185.1 1846 Silent Mutation CTC,CTG L162L NP_001309114.1
NM_001322186.1 1846 Silent Mutation CTC,CTG L410L NP_001309115.1
NM_001922.4 1846 Silent Mutation CTC,CTG L473L NP_001913.2
XM_011521049.1 1846 Silent Mutation CTC,CTG L410L XP_011519351.1
XM_017020401.1 1846 Silent Mutation CTC,CTG L516L XP_016875890.1

View Full Product Details