Product Details

SNP ID
rs150831702
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:48985050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTAAAAGTTAATTTAAAAACT[C/T]GTTGGAGTAGAGAAGGCCTTTTAAG
Phenotype
MIM: 615794
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNDC3A PubMed Links

Gene Details

Gene
FNDC3A
Gene Name
fibronectin type III domain containing 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079673.1 Intron NP_001073141.1
NM_001278438.1 Intron NP_001265367.1
NM_014923.4 Intron NP_055738.3
XM_011534997.2 Intron XP_011533299.1
XM_017020440.1 Intron XP_016875929.1
XM_017020441.1 Intron XP_016875930.1
XM_017020442.1 Intron XP_016875931.1
XM_017020443.1 Intron XP_016875932.1

View Full Product Details