Product Details

SNP ID
rs150898512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:106493172 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCTGTCCGCAGTCCTTAGCGGGAT[A/G]ATAATGTCACTGGGCTCTGAGCCGT
Phenotype
MIM: 600527
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFNB2 PubMed Links

Gene Details

Gene
EFNB2
Gene Name
ephrin B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004093.3 946 Silent Mutation ATC,ATT I290I NP_004084.1
XM_017020406.1 946 Silent Mutation ATC,ATT I292I XP_016875895.1

View Full Product Details