Product Details
- SNP ID
-
rs138709720
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:38209524 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCATGGAGGAGCCAGGGCGAAATGC[G/T]TCCCAGAACGGGACCTTGAGCGAGG
- Phenotype
-
MIM: 182451
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SSTR1
PubMed Links
Gene Details
- Gene
- SSTR1
- Gene Name
- somatostatin receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001049.2 |
752 |
Silent Mutation |
GCG,GCT |
A45A |
NP_001040.1 |
View Full Product Details