Product Details

SNP ID
rs139409894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20223944 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGCGGCTCCACACACCCATGTAC[A/C]TCCTTCTGGGAAACTTTGCCTTTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR11H6 PubMed Links

Gene Details

Gene
OR11H6
Gene Name
olfactory receptor family 11 subfamily H member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004480.1 235 Missense Mutation ATC,CTC I79L NP_001004480.1

View Full Product Details