Product Details

SNP ID
rs140064466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19920266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAACCTATGAGGCTTGTGGAGCTG[G/T]ACTCTAGGTCACTCCTCTAGTCTTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4K1 PubMed Links

Gene Details

Gene
OR4K1
Gene Name
olfactory receptor family 4 subfamily K member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004063.2 4 Intron NP_001004063.2
XM_011537153.2 4 UTR 5 XP_011535455.1
Gene
OR4K5
Gene Name
olfactory receptor family 4 subfamily K member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005483.1 4 Intron NP_001005483.1

View Full Product Details