Product Details

SNP ID
rs140455616
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.14:100538004 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTTCCGTAGAGCTGGGCCTTGG[A/T]GAGGCTGTCCTTGCGGCTCAGCCCC
Phenotype
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
BEGAIN PubMed Links

Gene Details

Gene
BEGAIN
Gene Name
brain enriched guanylate kinase associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159531.1 2035 Missense Mutation ACC,TCC T583S NP_001153003.1
NM_020836.3 2035 Missense Mutation ACC,TCC T583S NP_065887.1
XM_005267919.3 2035 Missense Mutation ACC,TCC T596S XP_005267976.1
XM_005267920.4 2035 Missense Mutation ACC,TCC T595S XP_005267977.1
XM_005267921.3 2035 Missense Mutation ACC,TCC T583S XP_005267978.1
XM_011537031.1 2035 Missense Mutation ACC,TCC T602S XP_011535333.1
XM_017021521.1 2035 Missense Mutation ACC,TCC T583S XP_016877010.1
XM_017021522.1 2035 Missense Mutation ACC,TCC T574S XP_016877011.1
Gene
WDR25
Gene Name
WD repeat domain 25
There are no transcripts associated with this gene.

View Full Product Details