Product Details

SNP ID
rs141833524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:21354516 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTCCAGTTGAGGGACTGTACTCC[C/T]TCTGTGTATTTCAGGTCGCAGGAAC
Phenotype
MIM: 605446 MIM: 605012
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPGRIP1 PubMed Links

Gene Details

Gene
RPGRIP1
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
There are no transcripts associated with this gene.

Gene
SUPT16H
Gene Name
SPT16 homolog, facilitates chromatin remodeling subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007192.3 3024 Silent Mutation GAA,GAG E895E NP_009123.1
XM_011536381.2 3024 Intron XP_011534683.1

View Full Product Details