Product Details

SNP ID
rs141999894
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24207162 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGTAGGGACTTAAAGTTGGAG[A/C]ACCTGTTGCTGGACAAGTGGGAGAA
Phenotype
MIM: 610051 MIM: 603171 MIM: 610711
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHMP4A PubMed Links

Gene Details

Gene
CHMP4A
Gene Name
charged multivesicular body protein 4A
There are no transcripts associated with this gene.

Gene
MDP1
Gene Name
magnesium dependent phosphatase 1
There are no transcripts associated with this gene.

Gene
NEDD8
Gene Name
neural precursor cell expressed, developmentally down-regulated 8
There are no transcripts associated with this gene.

Gene
NEDD8-MDP1
Gene Name
NEDD8-MDP1 readthrough
There are no transcripts associated with this gene.

Gene
TSSK4
Gene Name
testis specific serine kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184739.1 691 Missense Mutation AAC,CAC N163H NP_001171668.1
NM_001308067.1 691 Missense Mutation AAC,CAC N77H NP_001294996.1
NM_174944.3 691 Missense Mutation AAC,CAC N153H NP_777604.2
XM_011536663.2 691 Missense Mutation AAC,CAC N87H XP_011534965.1
XM_017021226.1 691 Missense Mutation AAC,CAC N153H XP_016876715.1

View Full Product Details