Product Details
- SNP ID
-
rs142089300
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:36477001 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCGGATGAACTCCTGCTTTGTGA[C/T]AGCGTCGTGGGATGCAGCAGGGCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SFTA3
PubMed Links
Gene Details
- Gene
- SFTA3
- Gene Name
- surfactant associated 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001101341.1 |
863 |
Silent Mutation |
CTA,CTG |
L77L |
NP_001094811.1 |
View Full Product Details