Product Details

SNP ID
rs142940048
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24306063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCTCGAGGGTTTTGCTTGTACAC[A/G]TCAAAGGTGAATCGGGCGATGTCCT
Phenotype
MIM: 604441 MIM: 610410 MIM: 601531 MIM: 605773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CIDEB PubMed Links

Gene Details

Gene
CIDEB
Gene Name
cell death-inducing DFFA-like effector b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318807.1 1547 Silent Mutation NP_001305736.1
NM_014430.2 1547 Silent Mutation NP_055245.2
XM_011536659.2 1547 Silent Mutation XP_011534961.1
XM_017021221.1 1547 Silent Mutation XP_016876710.1
Gene
DHRS1
Gene Name
dehydrogenase/reductase 1
There are no transcripts associated with this gene.

Gene
LTB4R
Gene Name
leukotriene B4 receptor
There are no transcripts associated with this gene.

Gene
LTB4R2
Gene Name
leukotriene B4 receptor 2
There are no transcripts associated with this gene.

Gene
NOP9
Gene Name
NOP9 nucleolar protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286367.1 1547 UTR 3 NP_001273296.1
NM_174913.2 1547 UTR 3 NP_777573.1
XM_005267385.1 1547 UTR 3 XP_005267442.1
XM_011536526.1 1547 UTR 3 XP_011534828.1
XM_011536527.2 1547 Intron XP_011534829.1

View Full Product Details