Product Details

SNP ID
rs142987949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75959217 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACATTTACAAGACTTCACCACCAT[A/G]TTGGAGAGCTGCTCCACTTTGGGGG
Phenotype
MIM: 190230 MIM: 612268
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TGFB3 PubMed Links

Gene Details

Gene
TGFB3
Gene Name
transforming growth factor beta 3
There are no transcripts associated with this gene.

Gene
TTLL5
Gene Name
tubulin tyrosine ligase like 5
There are no transcripts associated with this gene.

View Full Product Details