Product Details

SNP ID
rs143562391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:31106611 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACAAAAGCTTTAAAACTTATTCA[C/T]GCATTGGATTGAAGAATGTACATAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HECTD1 PubMed Links

Gene Details

Gene
HECTD1
Gene Name
HECT domain E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015382.3 Intron NP_056197.3
XM_005267502.2 Intron XP_005267559.2
XM_011536619.2 Intron XP_011534921.1
XM_011536621.2 Intron XP_011534923.1
XM_011536622.1 Intron XP_011534924.1
XM_017021148.1 Intron XP_016876637.1
XM_017021149.1 Intron XP_016876638.1
XM_017021150.1 Intron XP_016876639.1
XM_017021151.1 Intron XP_016876640.1
XM_017021152.1 Intron XP_016876641.1
XM_017021153.1 Intron XP_016876642.1

View Full Product Details