Product Details
- SNP ID
-
rs143803246
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:24574789 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTCCCGTCTCTGGATATTGTGGGC[G/T]CCCAGGGTGACATTTATATTGCTGC
- Phenotype
-
MIM: 116830
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CTSG
PubMed Links
Gene Details
- Gene
- CTSG
- Gene Name
- cathepsin G
View Full Product Details