Product Details

SNP ID
rs143876579
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20014656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTTGATCACCAGAGGGAGGTCAC[A/T]GAAGAAGCTGTCTACCTCATTGGGG
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR4K14 PubMed Links

Gene Details

Gene
OR4K14
Gene Name
olfactory receptor family 4 subfamily K member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004712.1 538 Missense Mutation AGT,TGT S180C NP_001004712.1

View Full Product Details