Product Details

SNP ID
rs143912363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24206554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGTACCTCATCAACTTCTATCGG[A/G]CCATTGAGAGCACATCTCGAGTATA
Phenotype
MIM: 610051 MIM: 610711
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHMP4A PubMed Links

Gene Details

Gene
CHMP4A
Gene Name
charged multivesicular body protein 4A
There are no transcripts associated with this gene.

Gene
MDP1
Gene Name
magnesium dependent phosphatase 1
There are no transcripts associated with this gene.

Gene
NEDD8-MDP1
Gene Name
NEDD8-MDP1 readthrough
There are no transcripts associated with this gene.

Gene
TSSK4
Gene Name
testis specific serine kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184739.1 475 Missense Mutation ACC,GCC T91A NP_001171668.1
NM_001308067.1 475 Missense Mutation ACC,GCC T15A NP_001294996.1
NM_174944.3 475 Missense Mutation ACC,GCC T91A NP_777604.2
XM_011536663.2 475 Missense Mutation ACC,GCC T15A XP_011534965.1
XM_017021226.1 475 Missense Mutation ACC,GCC T91A XP_016876715.1

View Full Product Details