Product Details

SNP ID
rs144919020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:55369702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCATCCCACCTGCACTGCTGCTCG[C/T]GATGGGTGGGGACGCCTGGGTGCTC
Phenotype
MIM: 613515
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATG14 PubMed Links

Gene Details

Gene
ATG14
Gene Name
autophagy related 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014924.4 1136 Missense Mutation NP_055739.2
XM_011536563.2 1136 Missense Mutation XP_011534865.1

View Full Product Details