Product Details
- SNP ID
-
rs145767522
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20770573 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGATAGCATAGAAGACCTAAAGAT[A/G]GTAGAACCTATCGGCAACTAGAAAG
- Phenotype
-
MIM: 611582
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EDDM3B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3748339] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EDDM3B
- Gene Name
- epididymal protein 3B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022360.4 |
521 |
Missense Mutation |
ATA,ATG |
I141M |
NP_071755.1 |
View Full Product Details