Product Details

SNP ID
rs145949964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23299585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAGATGAACATGAATGAAGAAG[A/G]AAACAAATGGAAATAAACAAAATGA
Phenotype
MIM: 601931
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL2L2 PubMed Links

Gene Details

Gene
BCL2L2
Gene Name
BCL2 like 2
There are no transcripts associated with this gene.

Gene
BCL2L2-PABPN1
Gene Name
BCL2L2-PABPN1 readthrough
There are no transcripts associated with this gene.

Gene
H3F3AP2
Gene Name
H3 histone, family 3A pseudogene 2
There are no transcripts associated with this gene.

Gene
PPP1R3E
Gene Name
protein phosphatase 1 regulatory subunit 3E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276318.1 Intron NP_001263247.1

View Full Product Details