Product Details

SNP ID
rs146377485
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:91969137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGATAATATTGCACAGGACCAAA[C/T]ACATACTTCACCTCAGGGAACCTAT
Phenotype
MIM: 604505
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIP11 PubMed Links

Gene Details

Gene
TRIP11
Gene Name
thyroid hormone receptor interactor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321851.1 6871 UTR 3 NP_001308780.1
NM_004239.4 6871 UTR 3 NP_004230.2
XM_017021787.1 6871 UTR 3 XP_016877276.1
XM_017021788.1 6871 UTR 3 XP_016877277.1

View Full Product Details