Product Details

SNP ID
rs147817006
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:64550002 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGGCTTCCAGGGCGAGGCCATGT[A/G]CCGGGTGCCCGAGTTTTATGCGAGG
Phenotype
MIM: 140560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPA2 PubMed Links

Gene Details

Gene
HSPA2
Gene Name
heat shock protein family A (Hsp70) member 2
There are no transcripts associated with this gene.

Gene
LOC102723809
Gene Name
uncharacterized LOC102723809
There are no transcripts associated with this gene.

Gene
PPP1R36
Gene Name
protein phosphatase 1 regulatory subunit 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172365.2 69 Missense Mutation TAC,TGC Y2C NP_758953.1
XM_005267354.3 69 Intron XP_005267411.1
XM_005267355.3 69 UTR 5 XP_005267412.1
XM_011536468.2 69 Missense Mutation TAC,TGC Y2C XP_011534770.1

View Full Product Details