Product Details

SNP ID
rs148072819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24115650 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGGTCCGGAGACCCCTCCGAGG[A/G]CTTGCCCCGAAGAGGGGCTGGCCTG
Phenotype
MIM: 613317 MIM: 162080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF11 PubMed Links

Gene Details

Gene
DCAF11
Gene Name
DDB1 and CUL4 associated factor 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163484.1 786 Missense Mutation GAC,GGC D19G NP_001156956.1
NM_025230.4 786 Missense Mutation GAC,GGC D19G NP_079506.3
NM_181357.2 786 Missense Mutation GAC,GGC D19G NP_852002.1
Gene
NRL
Gene Name
neural retina leucine zipper
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006177.3 786 Intron NP_006168.1
XM_005267708.4 786 Intron XP_005267765.1
XM_005267709.3 786 Intron XP_005267766.1
XM_005267710.3 786 Intron XP_005267767.1
XM_011536801.2 786 Intron XP_011535103.2
XM_011536802.1 786 Intron XP_011535104.1
XM_011536804.2 786 Intron XP_011535106.1
XM_011536805.2 786 Intron XP_011535107.1
XM_011536806.2 786 Intron XP_011535108.2
XM_017021350.1 786 Intron XP_016876839.1

View Full Product Details