Product Details

SNP ID
rs148996083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24164652 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCACCTTCATCTACAACGGGCGC[A/G]TGGTGGGCGAGGCCCAGGTGCAAAG
Phenotype
MIM: 147574 MIM: 608193 MIM: 612487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRF9 PubMed Links

Gene Details

Gene
IRF9
Gene Name
interferon regulatory factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006084.4 815 Missense Mutation ATG,GTG M230V NP_006075.3
Gene
REC8
Gene Name
REC8 meiotic recombination protein
There are no transcripts associated with this gene.

Gene
RNF31
Gene Name
ring finger protein 31
There are no transcripts associated with this gene.

View Full Product Details