Product Details

SNP ID
rs149838792
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:20014632 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGACATAGGTGTCCATGCAGGCAA[A/G]TTTGATCACCAGAGGGAGGTCACAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR4K14 PubMed Links

Gene Details

Gene
OR4K14
Gene Name
olfactory receptor family 4 subfamily K member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004712.1 562 Missense Mutation CTT,TTT L188F NP_001004712.1

View Full Product Details